Alfa Aesar™ 2-Amino-3-bromo-5-chloropyridine, 95%

Manufacturer: Alfa Aesar™ H2779903

Quantity 1g

 View more versions of this product

Catalog No. AAH2779903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26163-03-1
    Molecular Formula C5H4BrClN2
    Formula Weight 207.46
    MDL Number MFCD00955627
    Synonym 3-Bromo-5-chloro-2-pyridinamine
    Chemical Name or Material 2-Amino-3-bromo-5-chloropyridine

    Specifications

    Formula Weight 207.46
    Melting Point 83° to 85°C
    Molecular Formula C5H4BrClN2
    Quantity 1g
    Synonym 3-Bromo-5-chloro-2-pyridinamine
    MDL Number MFCD00955627
    CAS 26163-03-1
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Amino-3-bromo-5-chloropyridine
    Assay 95%

    GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit