Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-3-bromo-5-fluorobenzoic acid, 95%

Click to view available options
Quantity:
250mg
1g
259269-84-6
C7H5BrFNO2
234.024
MFCD00662509
LTOCPZGBNWZMBA-UHFFFAOYSA-N
benzoic acid, 2-amino-3-bromo-5-fluoro,2-amino-3-bromo-5-fluoro-benzoic acid,2-amino-3-bromo-5-fluorobenzoicacid
2781703
2-amino-3-bromo-5-fluorobenzoic acid
C1=C(C=C(C(=C1Br)N)C(=O)O)F
259269-84-6
C7H5BrFNO2
234.024
MFCD00662509
LTOCPZGBNWZMBA-UHFFFAOYSA-N
benzoic acid, 2-amino-3-bromo-5-fluoro,2-amino-3-bromo-5-fluoro-benzoic acid,2-amino-3-bromo-5-fluorobenzoicacid
2781703
2-amino-3-bromo-5-fluorobenzoic acid
C1=C(C=C(C(=C1Br)N)C(=O)O)F
Quantity 1g
Beilstein 15583945
Formula Weight 234.02
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Amino-3-bromo-5-fluorobenzoic acid