Alfa Aesar™ 2-Amino-3-bromo-5-nitrobenzonitrile, 98%

Manufacturer: Alfa Aesar™ B2540406

Quantity 5g

 View more versions of this product

Catalog No. AAB2540406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 17601-94-4
    Molecular Formula C7H4BrN3O2
    Formula Weight 242.03
    MDL Number MFCD00054185
    UN Number UN3439
    Synonym 2-Bromo-6-cyano-4-nitroaniline
    Chemical Name or Material 2-Amino-3-bromo-5-nitrobenzonitrile

    Specifications

    Formula Weight 242.03
    Melting Point 186° to 190°C
    Quantity 5g
    TSCA TSCA
    Molecular Formula C7H4BrN3O2
    MDL Number MFCD00054185
    UN Number UN3439
    Synonym 2-Bromo-6-cyano-4-nitroaniline
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P307+P311
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed: Call a POISON CENTER or doctor/physician.
    CAS 17601-94-4
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H312-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 2-Amino-3-bromo-5-nitrobenzonitrile
    Assay 98%

    GHS H Statement
    H301-H312-H332-H315-H319-H335
    Toxic if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P307+P311
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed: Call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit