Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-3-bromo-6-methylpyridine, 95%

Click to view available options
Quantity:
1g
5g
126325-46-0
C6H7BrN2
187.04
MFCD03095215
JYWWGZAAXTYNRN-UHFFFAOYSA-N
2-amino-3-bromo-6-methylpyridine,2-amino-3-bromo-6-picoline,6-amino-5-bromo-2-picoline,2-pyridinamine, 3-bromo-6-methyl,3-bromo-6-methyl-2-pyridylamine,2-amine-3-bromo-6-methylpyridine,3-bromo-6-methyl-pyridin-2-amine,2-amino-3-bromo-6-picoline 2-amino-3-bromo-6-methylpyridine,pubchem1235,6-amino-5-bromo-picoline
2734416
3-bromo-6-methylpyridin-2-amine
CC1=NC(=C(C=C1)Br)N
126325-46-0
C6H7BrN2
187.04
MFCD03095215
JYWWGZAAXTYNRN-UHFFFAOYSA-N
2-amino-3-bromo-6-methylpyridine,2-amino-3-bromo-6-picoline,6-amino-5-bromo-2-picoline,2-pyridinamine, 3-bromo-6-methyl,3-bromo-6-methyl-2-pyridylamine,2-amine-3-bromo-6-methylpyridine,3-bromo-6-methyl-pyridin-2-amine,2-amino-3-bromo-6-picoline 2-amino-3-bromo-6-methylpyridine,pubchem1235,6-amino-5-bromo-picoline
2734416
3-bromo-6-methylpyridin-2-amine
CC1=NC(=C(C=C1)Br)N
Quantity 1g
Color White
Sensitivity Light Sensitive
Melting Point 78°C to 83°C
UN Number UN2811
Beilstein 6190026
Formula Weight 187.04
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Amino-3-bromo-6-methylpyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C