Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-bromopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13534-99-1
C5H5BrN2
173.01
MFCD03095187
RBCARPJOEUEZLS-UHFFFAOYSA-N
2-amino-3-bromopyridine,3-bromo-2-pyridinamine,2-pyridinamine, 3-bromo,3-bromo-2-pyridylamine,3-bromo-pyridin-2-ylamine,3-bromanylpyridin-2-amine,bromo-aminopyridine,bromopyridin-2-amine,pubchem1241,3-bromo-2-aminopyridine
817700
3-bromopyridin-2-amine
C1=CC(=C(N=C1)N)Br
13534-99-1
C5H5BrN2
173.01
MFCD03095187
RBCARPJOEUEZLS-UHFFFAOYSA-N
2-amino-3-bromopyridine,3-bromo-2-pyridinamine,2-pyridinamine, 3-bromo,3-bromo-2-pyridylamine,3-bromo-pyridin-2-ylamine,3-bromanylpyridin-2-amine,bromo-aminopyridine,bromopyridin-2-amine,pubchem1241,3-bromo-2-aminopyridine
817700
3-bromopyridin-2-amine
C1=CC(=C(N=C1)N)Br
Quantity 25g
Melting Point 61.0°C to 65.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown to Gray
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 173.01
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-3-bromopyridine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning