Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-chlorobenzotrifluoride 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
433-94-3
C7H5ClF3N
195.569
MFCD00272565
OTRRSPQJZRCMDA-UHFFFAOYSA-N
2-chloro-6-trifluoromethyl aniline,2-amino-3-chlorobenzotrifluoride,benzenamine, 2-chloro-6-trifluoromethyl,2-chloranyl-6-trifluoromethyl aniline,pubchem2764,2-chloro-6-trifluoromethyl benzenamine,2-chloro-6-trifluoromethyl phenylamine,6-chloro-2-trifluoromethyl phenylamine,2-chloro-6-trifluoromethyl-benzenamine,2-chloro-6-trifluoromethyl aniline 2-amino-3-chlorobenzotrifluoride
2756384
2-chloro-6-(trifluoromethyl)aniline
C1=CC(=C(C(=C1)Cl)N)C(F)(F)F
433-94-3
C7H5ClF3N
195.569
MFCD00272565
OTRRSPQJZRCMDA-UHFFFAOYSA-N
2-chloro-6-trifluoromethyl aniline,2-amino-3-chlorobenzotrifluoride,benzenamine, 2-chloro-6-trifluoromethyl,2-chloranyl-6-trifluoromethyl aniline,pubchem2764,2-chloro-6-trifluoromethyl benzenamine,2-chloro-6-trifluoromethyl phenylamine,6-chloro-2-trifluoromethyl phenylamine,2-chloro-6-trifluoromethyl-benzenamine,2-chloro-6-trifluoromethyl aniline 2-amino-3-chlorobenzotrifluoride
2756384
2-chloro-6-(trifluoromethyl)aniline
C1=CC(=C(C(=C1)Cl)N)C(F)(F)F
Boiling Point 40°C
Color Yellow
Quantity 25g
Formula Weight 195.57
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material 2-Amino-3-chlorobenzotrifluoride

EINECSNumber : (3)-0266

TSCA : No