Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-hydroxypyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
16867-03-1
C5H6N2O
110.12
MFCD00006317
BMTSZVZQNMNPCT-UHFFFAOYSA-N
2-amino-3-hydroxypyridine,2-amino-3-pyridinol,3-pyridinol, 2-amino,2-amino-3-pyridol,3-hydroxy-2-pyridinamine,2-amino-pyridin-3-ol,2-amino-3-hydroxy pyridine,unii-p8740pew4b,3-hydroxy-2-aminopyridine,pubchem2574
28114
2-aminopyridin-3-ol
C1=CC(=C(N=C1)N)O
16867-03-1
C5H6N2O
110.12
MFCD00006317
BMTSZVZQNMNPCT-UHFFFAOYSA-N
2-amino-3-hydroxypyridine,2-amino-3-pyridinol,3-pyridinol, 2-amino,2-amino-3-pyridol,3-hydroxy-2-pyridinamine,2-amino-pyridin-3-ol,2-amino-3-hydroxy pyridine,unii-p8740pew4b,3-hydroxy-2-aminopyridine,pubchem2574
28114
2-aminopyridin-3-ol
C1=CC(=C(N=C1)N)O
Color Beige-Gray to Brown
Packaging Glass bottle
Water 0.5% max. (K.F.)
Infrared Spectrum Authentic
Quantity 25g
Melting Point 170°C to 176°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula HO(C5H3N)NH2
Beilstein 22, II, 408
Formula Weight 110.12
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-3-hydroxypyridine, 98%
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 240-886-8

TSCA : TSCA