Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-iodopyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
104830-06-0
C5H5IN2
220.01
UUDNBWSHTUFGDQ-UHFFFAOYSA-N
2-amino-3-iodopyridine,3-iodopyrid-2-ylamine,3-iodo-pyridin-2-ylamine,3-iodo-2-pyridylamine,2-pyridinamine, 3-iodo,iodo-aminopyridine,pubchem1265,2-amino-3-iodpyridine,acmc-20a0df
2763151
3-iodopyridin-2-amine
C1=CC(=C(N=C1)N)I
104830-06-0
C5H5IN2
220.01
UUDNBWSHTUFGDQ-UHFFFAOYSA-N
2-amino-3-iodopyridine,3-iodopyrid-2-ylamine,3-iodo-pyridin-2-ylamine,3-iodo-2-pyridylamine,2-pyridinamine, 3-iodo,iodo-aminopyridine,pubchem1265,2-amino-3-iodpyridine,acmc-20a0df
2763151
3-iodopyridin-2-amine
C1=CC(=C(N=C1)N)I
Color Brown to White
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 89.6°C to 90.6°C
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 220.01
Physical Form Powder, Crystalline Powder or Needles
Percent Purity 97%
Chemical Name or Material 2-Amino-3-iodopyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning