Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-methoxybenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
3177-80-8
C8H9NO3
167.17
SXOPCLUOUFQBJV-UHFFFAOYSA-N
3-methoxyanthranilic acid,benzoic acid, 2-amino-3-methoxy,3-methoxyanthranilate,m-anisic acid, 2-amino,3-methoxy-2-aminobenzoic acid,2-amino-3-methoxy-benzoic acid,2-amino-3-methoxy benzoic acid,2-amino-3-methyloxy benzoic acid,2-amino-3-methoxybenzoicacid
255720
CHEBI:27440
2-amino-3-methoxybenzoic acid
COC1=CC=CC(=C1N)C(=O)O
3177-80-8
C8H9NO3
167.17
SXOPCLUOUFQBJV-UHFFFAOYSA-N
3-methoxyanthranilic acid,benzoic acid, 2-amino-3-methoxy,3-methoxyanthranilate,m-anisic acid, 2-amino,3-methoxy-2-aminobenzoic acid,2-amino-3-methoxy-benzoic acid,2-amino-3-methoxy benzoic acid,2-amino-3-methyloxy benzoic acid,2-amino-3-methoxybenzoicacid
255720
CHEBI:27440
2-amino-3-methoxybenzoic acid
COC1=CC=CC(=C1N)C(=O)O
Melting Point 168°C to 170.7°C
Color Brown to White
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 167.17
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-3-methoxybenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning