Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-methoxypyridine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
10201-71-5
C6H8N2O
124.14
HNAYRVKSWGSQTP-UHFFFAOYSA-N
2-amino-3-methoxypyridine,2-pyridinamine, 3-methoxy,3-methoxy-2-pyridylamine,3-methoxy-pyridin-2-ylamine,3-methoxy-2-pyridinamine,acmc-1c0wi,3-methoxypyridine-2-amine,3-methoxy-2-aminopyridine,3-methoxy-pyridin-2-amine,ksc494m4p
9920384
3-methoxypyridin-2-amine
COC1=C(N=CC=C1)N
10201-71-5
C6H8N2O
124.14
HNAYRVKSWGSQTP-UHFFFAOYSA-N
2-amino-3-methoxypyridine,2-pyridinamine, 3-methoxy,3-methoxy-2-pyridylamine,3-methoxy-pyridin-2-ylamine,3-methoxy-2-pyridinamine,acmc-1c0wi,3-methoxypyridine-2-amine,3-methoxy-2-aminopyridine,3-methoxy-pyridin-2-amine,ksc494m4p
9920384
3-methoxypyridin-2-amine
COC1=C(N=CC=C1)N
Melting Point 75°C to 77°C
Packaging Glass Bottle
Quantity 1g
Assay Percent Range 96% min. (GC)
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-methoxypyridine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning