Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-nitrophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
603-85-0
C6H6N2O3
154.12
MFCD00010875
KUCWUAFNGCMZDB-UHFFFAOYSA-N
phenol, 2-amino-3-nitro,2-hydroxy-6-nitroaniline,ccris 2544,unii-a7f97zpu2w,3-nitro-2-aminophenol,2-amino-3-nitro-phenol,a7f97zpu2w,2-amino-3-hydroxynitrobenzene,1-hydroxy-2-amino-3-nitrobenzene,2-amino-nitrophenol
4115495
2-amino-3-nitrophenol
C1=CC(=C(C(=C1)O)N)[N+](=O)[O-]
603-85-0
C6H6N2O3
154.12
MFCD00010875
KUCWUAFNGCMZDB-UHFFFAOYSA-N
phenol, 2-amino-3-nitro,2-hydroxy-6-nitroaniline,ccris 2544,unii-a7f97zpu2w,3-nitro-2-aminophenol,2-amino-3-nitro-phenol,a7f97zpu2w,2-amino-3-hydroxynitrobenzene,1-hydroxy-2-amino-3-nitrobenzene,2-amino-nitrophenol
4115495
2-amino-3-nitrophenol
C1=CC(=C(C(=C1)O)N)[N+](=O)[O-]
Infrared Spectrum Authentic
Packaging Glass bottle
Color Red to Brown
Quantity 5g
Melting Point 215.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula H2NC6H3(NO2)OH
Solubility Information Solubility in water: slightly soluble.
Formula Weight 154.12
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-3-nitrophenol
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective g

GHS Signal Word: Warning

EINECSNumber : 210-060-1