Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-nitrophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
603-85-0
C6H6N2O3
154.12
MFCD00010875
KUCWUAFNGCMZDB-UHFFFAOYSA-N
phenol, 2-amino-3-nitro,2-hydroxy-6-nitroaniline,ccris 2544,unii-a7f97zpu2w,3-nitro-2-aminophenol,2-amino-3-nitro-phenol,a7f97zpu2w,2-amino-3-hydroxynitrobenzene,1-hydroxy-2-amino-3-nitrobenzene,2-amino-nitrophenol
4115495
2-amino-3-nitrophenol
C1=CC(=C(C(=C1)O)N)[N+](=O)[O-]
603-85-0
C6H6N2O3
154.12
MFCD00010875
KUCWUAFNGCMZDB-UHFFFAOYSA-N
phenol, 2-amino-3-nitro,2-hydroxy-6-nitroaniline,ccris 2544,unii-a7f97zpu2w,3-nitro-2-aminophenol,2-amino-3-nitro-phenol,a7f97zpu2w,2-amino-3-hydroxynitrobenzene,1-hydroxy-2-amino-3-nitrobenzene,2-amino-nitrophenol
4115495
2-amino-3-nitrophenol
C1=CC(=C(C(=C1)O)N)[N+](=O)[O-]
Melting Point 215.0°C
Quantity 25g
Packaging Glass bottle
Color Red to Brown
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula H2NC6H3(NO2)OH
Solubility Information Solubility in water: slightly soluble.
Formula Weight 154.12
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-3-nitrophenol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.