Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-nitropyridine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4214-75-9
C5H5N3O2
139.11
MFCD00006314
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
2-amino-3-nitropyridine,3-nitro-2-aminopyridine,2-pyridinamine, 3-nitro,3-nitropyridin-2-ylamine,3-nitro-pyridin-2-ylamine,pyridine, 2-amino-3-nitro,2-amino3-nitropyridine,3-nitro-2-pyridinamine,2-amino-3-nitro pyridine,3-nitro-2-pyridylamine
77887
3-nitropyridin-2-amine
C1=CC(=C(N=C1)N)[N+](=O)[O-]
4214-75-9
C5H5N3O2
139.11
MFCD00006314
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
2-amino-3-nitropyridine,3-nitro-2-aminopyridine,2-pyridinamine, 3-nitro,3-nitropyridin-2-ylamine,3-nitro-pyridin-2-ylamine,pyridine, 2-amino-3-nitro,2-amino3-nitropyridine,3-nitro-2-pyridinamine,2-amino-3-nitro pyridine,3-nitro-2-pyridylamine
77887
3-nitropyridin-2-amine
C1=CC(=C(N=C1)N)[N+](=O)[O-]
Flash Point 167°C
Melting Point 165.9°C to 166.4°C
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula NH2(C5H3N)NO2
Beilstein 22, I, 631
Solubility Information Solubility in water: 3g/L (20 c)
Formula Weight 139.11
Physical Form Crystalline Powder and Needles
Percent Purity 99%
Chemical Name or Material 2-Amino-3-nitropyridine, 99%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 224-144-0

TSCA : TSCA