Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-amino-3-pyridinecarboxaldehyde 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7521-41-7
C6H6N2O
122.127
MFCD01830382
NXMFJCRMSDRXLD-UHFFFAOYSA-N
2-aminonicotinaldehyde,2-amino-3-pyridinecarboxaldehyde,2-amino-3-formylpyridine,3-pyridinecarboxaldehyde, 2-amino,2-amino-pyridine-3-carbaldehyde,2-aminopyridine-3-carboxaldehyde,2-amino-3-pyridine carboxaldehyde,2-amino-pyridin-3-carbaldehyde,pubchem1262,2-amino nicotinaldehyde
737633
2-aminopyridine-3-carbaldehyde
C1=CC(=C(N=C1)N)C=O
7521-41-7
C6H6N2O
122.127
MFCD01830382
NXMFJCRMSDRXLD-UHFFFAOYSA-N
2-aminonicotinaldehyde,2-amino-3-pyridinecarboxaldehyde,2-amino-3-formylpyridine,3-pyridinecarboxaldehyde, 2-amino,2-amino-pyridine-3-carbaldehyde,2-aminopyridine-3-carboxaldehyde,2-amino-3-pyridine carboxaldehyde,2-amino-pyridin-3-carbaldehyde,pubchem1262,2-amino nicotinaldehyde
737633
2-aminopyridine-3-carbaldehyde
C1=CC(=C(N=C1)N)C=O
Infrared Spectrum Authentic
Color Yellow to Brown
Quantity 5g
Packaging Glass bottle
Melting Point 95°C to 102°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 122.13
Physical Form Powder, Crystals and/or Chunks
Percent Purity ≥96.0%
Chemical Name or Material 2-Amino-3-pyridinecarboxaldehyde
Show More Show Less

GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning