Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-(1-naphthyl)thiazole, 97%

Click to view available options
Quantity:
1g
5g
56503-96-9
C13H10N2S
226.297
MFCD00236016
NBQUWOCIFFHZTM-UHFFFAOYSA-N
2-amino-4-1-naphthyl thiazole,4-1-naphthalenyl-2-thiazolamine,4-naphthalen-1-yl-thiazol-2-ylamine,2-thiazolamine, 4-1-naphthalenyl,4-naphthalen-1-yl-1,3-thiazol-2-amine,4-naphthalen-1-yl thiazol-2-amine,4-naphthalen-1-yl-thiazol-2-yl-amine,4-1-naphthyl-1,3-thiazol-2-amine,2-amino-4-alpha-naphthylthiazole,2-amino-4-1-naphthalenyl-1,3-thiazole
151342
4-naphthalen-1-yl-1,3-thiazol-2-amine
C1=CC=C2C(=C1)C=CC=C2C3=CSC(=N3)N
56503-96-9
C13H10N2S
226.297
MFCD00236016
NBQUWOCIFFHZTM-UHFFFAOYSA-N
2-amino-4-1-naphthyl thiazole,4-1-naphthalenyl-2-thiazolamine,4-naphthalen-1-yl-thiazol-2-ylamine,2-thiazolamine, 4-1-naphthalenyl,4-naphthalen-1-yl-1,3-thiazol-2-amine,4-naphthalen-1-yl thiazol-2-amine,4-naphthalen-1-yl-thiazol-2-yl-amine,4-1-naphthyl-1,3-thiazol-2-amine,2-amino-4-alpha-naphthylthiazole,2-amino-4-1-naphthalenyl-1,3-thiazole
151342
4-naphthalen-1-yl-1,3-thiazol-2-amine
C1=CC=C2C(=C1)C=CC=C2C3=CSC(=N3)N
Quantity 5g
Melting Point 155°C to 158°C
Beilstein 164858
Formula Weight 226.3
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(1-naphthyl)thiazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XJ1990000