Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-(2-naphthyl)thiazole, 98%

Click to view available options
Quantity:
1g
5g
21331-43-1
C13H10N2S
226.297
MFCD00046452
GWDNDNTTXIIXRS-UHFFFAOYSA-N
4-naphthalen-2-yl-thiazol-2-ylamine,4-naphthalen-2-yl-1,3-thiazol-2-amine,4-2-naphthyl-1,3-thiazol-2-amine,2-amino-4-2-naphthyl thiazole,4-naphthalen-2-yl thiazol-2-amine,4-naphthalen-2-yl-thiazol-2-yl-amine,4-2-naphthalenyl-2-thiazolamine,2-thiazolamine,4-2-naphthalenyl,4-2-naphthyl-1,3-thiazole-2-ylamine,acmc-20ao57
673702
4-naphthalen-2-yl-1,3-thiazol-2-amine
C1=CC=C2C=C(C=CC2=C1)C3=CSC(=N3)N
21331-43-1
C13H10N2S
226.297
MFCD00046452
GWDNDNTTXIIXRS-UHFFFAOYSA-N
4-naphthalen-2-yl-thiazol-2-ylamine,4-naphthalen-2-yl-1,3-thiazol-2-amine,4-2-naphthyl-1,3-thiazol-2-amine,2-amino-4-2-naphthyl thiazole,4-naphthalen-2-yl thiazol-2-amine,4-naphthalen-2-yl-thiazol-2-yl-amine,4-2-naphthalenyl-2-thiazolamine,2-thiazolamine,4-2-naphthalenyl,4-2-naphthyl-1,3-thiazole-2-ylamine,acmc-20ao57
673702
4-naphthalen-2-yl-1,3-thiazol-2-amine
C1=CC=C2C=C(C=CC2=C1)C3=CSC(=N3)N
Quantity 1g
Melting Point 153°C to 156°C
Beilstein 173533
Formula Weight 226.3
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(2-naphthyl)thiazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning