Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-(2-naphthyl)thiazole, 98%

Click to view available options
Quantity:
1g
5g
21331-43-1
C13H10N2S
226.297
MFCD00046452
GWDNDNTTXIIXRS-UHFFFAOYSA-N
4-naphthalen-2-yl-thiazol-2-ylamine,4-naphthalen-2-yl-1,3-thiazol-2-amine,4-2-naphthyl-1,3-thiazol-2-amine,2-amino-4-2-naphthyl thiazole,4-naphthalen-2-yl thiazol-2-amine,4-naphthalen-2-yl-thiazol-2-yl-amine,4-2-naphthalenyl-2-thiazolamine,2-thiazolamine,4-2-naphthalenyl,4-2-naphthyl-1,3-thiazole-2-ylamine,acmc-20ao57
673702
4-naphthalen-2-yl-1,3-thiazol-2-amine
C1=CC=C2C=C(C=CC2=C1)C3=CSC(=N3)N
21331-43-1
C13H10N2S
226.297
MFCD00046452
GWDNDNTTXIIXRS-UHFFFAOYSA-N
4-naphthalen-2-yl-thiazol-2-ylamine,4-naphthalen-2-yl-1,3-thiazol-2-amine,4-2-naphthyl-1,3-thiazol-2-amine,2-amino-4-2-naphthyl thiazole,4-naphthalen-2-yl thiazol-2-amine,4-naphthalen-2-yl-thiazol-2-yl-amine,4-2-naphthalenyl-2-thiazolamine,2-thiazolamine,4-2-naphthalenyl,4-2-naphthyl-1,3-thiazole-2-ylamine,acmc-20ao57
673702
4-naphthalen-2-yl-1,3-thiazol-2-amine
C1=CC=C2C=C(C=CC2=C1)C3=CSC(=N3)N
Melting Point 153°C to 156°C
Quantity 5g
Beilstein 173533
Formula Weight 226.3
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(2-naphthyl)thiazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

DOTInformation : Hazard Class: 9; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures