Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-(3,4-difluorophenyl)thiazole, 97%

Click to view available options
Quantity:
250mg
1g
175135-32-7
C9H6F2N2S
212.218
MFCD00052876
NDCSJUJQMRFHEX-UHFFFAOYSA-N
4-3,4-difluorophenyl-1,3-thiazol-2-amine,2-amino-4-3,4-difluorophenyl thiazole,4-3,4-difluorophenyl thiazol-2-amine,4-3,4-difluoro-phenyl-thiazol-2-ylamine,2-amino-4-3,4-difluorophenyl-1,3-thiazole,2-thiazolamine,4-3,4-difluorophenyl,4-3,4-difluoro-phenyl-1,3-thiazol-2-amine,4-3,4-difluorophenyl-1,3-thiazole-2-ylamine,pubchem14456,timtec-bb sbb005523
706326
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
C1=CC(=C(C=C1C2=CSC(=N2)N)F)F
175135-32-7
C9H6F2N2S
212.218
MFCD00052876
NDCSJUJQMRFHEX-UHFFFAOYSA-N
4-3,4-difluorophenyl-1,3-thiazol-2-amine,2-amino-4-3,4-difluorophenyl thiazole,4-3,4-difluorophenyl thiazol-2-amine,4-3,4-difluoro-phenyl-thiazol-2-ylamine,2-amino-4-3,4-difluorophenyl-1,3-thiazole,2-thiazolamine,4-3,4-difluorophenyl,4-3,4-difluoro-phenyl-1,3-thiazol-2-amine,4-3,4-difluorophenyl-1,3-thiazole-2-ylamine,pubchem14456,timtec-bb sbb005523
706326
4-(3,4-difluorophenyl)-1,3-thiazol-2-amine
C1=CC(=C(C=C1C2=CSC(=N2)N)F)F
Quantity 1g
Color Brown to White
Melting Point 121°C to 125°C
Formula Weight 212.22
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(3,4-difluorophenyl)thiazole

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning