Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxamide, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
4815-28-5
C9H12N2OS
196.27
FFAKFORHXDNYEN-UHFFFAOYSA-N
2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide,benzo b thiophene-3-carboxamide, 2-amino-4,5,6,7-tetrahydro,2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide,2-amino-3-carboxamido-4,5,6,7-tetrahydro benzo b thiophene,benzo b thiophene-3-carboxamide,2-amino-4,5,6,7-tetrahydro,2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxamide,2-amino-3-carbamoyl-4,5,6,7-tetrahydrobenzo b thiophene,2-amino-3-carboxamido-4,5,6,7-tetrahydrobenzo b thiophene,2-amino-4,5,6,7-tetrahydro-benzo b thiophen-3-carboxamid,2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carboxamide
290223
2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
C1CCC2=C(C1)C(=C(S2)N)C(=O)N
4815-28-5
C9H12N2OS
196.27
FFAKFORHXDNYEN-UHFFFAOYSA-N
2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide,benzo b thiophene-3-carboxamide, 2-amino-4,5,6,7-tetrahydro,2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide,2-amino-3-carboxamido-4,5,6,7-tetrahydro benzo b thiophene,benzo b thiophene-3-carboxamide,2-amino-4,5,6,7-tetrahydro,2-amino-4,5,6,7-tetrahydrobenzothiophene-3-carboxamide,2-amino-3-carbamoyl-4,5,6,7-tetrahydrobenzo b thiophene,2-amino-3-carboxamido-4,5,6,7-tetrahydrobenzo b thiophene,2-amino-4,5,6,7-tetrahydro-benzo b thiophen-3-carboxamid,2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carboxamide
290223
2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
C1CCC2=C(C1)C(=C(S2)N)C(=O)N
Melting Point 181°C to 183°C
Quantity 5g
Assay Percent Range 96% min. (HPLC)
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxamide

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning