Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4,5-diphenylthiazole, 97%

Manufacturer:  Alfa Aesar™ H5181803

Quantity 1g

 View more versions of this product

Catalog No. AAH5181803


  Check Availability
Add to cart

Specifications

Description

Applications
Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne).

Solubility
Insoluble in water. Soluble in chloroform and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

6318-74-7
252.33
4,5-Diphenyl-2-thiazolamine
C15H12N2S
MFCD00438803
2-Amino-4,5-diphenylthiazole

Specifications

6318-74-7
Odorless
252.33
4,5-Diphenyl-2-thiazolamine
97%
1g
C15H12N2S
MFCD00438803
2-Amino-4,5-diphenylthiazole

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : XJ2450000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit