Alfa Aesar™ 2-Amino-4,5-diphenylthiazole, 97%

Manufacturer: Alfa Aesar™ H5181806

Quantity 5g

 View more versions of this product

Catalog No. AAH5181806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6318-74-7
    Molecular Formula C15H12N2S
    Formula Weight 252.33
    MDL Number MFCD00438803
    Synonym 4,5-Diphenyl-2-thiazolamine
    Chemical Name or Material 2-Amino-4,5-diphenylthiazole

    Specifications

    Formula Weight 252.33
    Quantity 5g
    Odor Odorless
    MDL Number MFCD00438803
    Synonym 4,5-Diphenyl-2-thiazolamine
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C15H12N2S
    CAS 6318-74-7
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number XJ2450000
    Chemical Name or Material 2-Amino-4,5-diphenylthiazole
    Assay 97%

    Description

    Applications
    Bis(trimethylsilyl)acetylene is a ligand that binds to a central metal ion (e.g. Titanium) to create a coordination complex. One of its complexes, Permethyltitanocene-bis(trimethylsilyl)acetylene, is a catalyst for the head-to-tail dimerization of 1-alkynes (e.g. 3-Penten-1-yne).

    Solubility
    Insoluble in water. Soluble in chloroform and methanol.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit