Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride, 98%

Click to view available options
Quantity:
5g
25g
100g
93983-01-8
C9H10ClNO3
215.633
MFCD00053022
PIVDPBIZDWHINL-UHFFFAOYSA-N
2'-amino-4',5'-methylenedioxyacetophenone hydrochloride,1-6-aminobenzo d 1,3 dioxol-5-yl ethanone hydrochloride,1-6-amino-1,3-benzodioxol-5-yl ethan-1-one hydrochloride,2-amino-4,5-methylenedioxyacetophenone hydrochloride,1-6-amino-2h-1,3-benzodioxol-5-yl ethanone hydrochloride,1-6-amino-1,3-benzodioxol-5-yl ethanone hydrochloride,1-6-azanyl-1,3-benzodioxol-5-yl ethanone hydrochloride
16205927
1-(6-amino-1,3-benzodioxol-5-yl)ethanone;hydrochloride
CC(=O)C1=CC2=C(C=C1N)OCO2.Cl
93983-01-8
C9H10ClNO3
215.633
MFCD00053022
PIVDPBIZDWHINL-UHFFFAOYSA-N
2'-amino-4',5'-methylenedioxyacetophenone hydrochloride,1-6-aminobenzo d 1,3 dioxol-5-yl ethanone hydrochloride,1-6-amino-1,3-benzodioxol-5-yl ethan-1-one hydrochloride,2-amino-4,5-methylenedioxyacetophenone hydrochloride,1-6-amino-2h-1,3-benzodioxol-5-yl ethanone hydrochloride,1-6-amino-1,3-benzodioxol-5-yl ethanone hydrochloride,1-6-azanyl-1,3-benzodioxol-5-yl ethanone hydrochloride
16205927
1-(6-amino-1,3-benzodioxol-5-yl)ethanone;hydrochloride
CC(=O)C1=CC2=C(C=C1N)OCO2.Cl
Quantity 100g
Formula Weight 215.64
Percent Purity 98%
Chemical Name or Material 2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning