Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,6-dichlorophenol 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
527-62-8
C6H5Cl2NO
178.012
MFCD00035766
WASQBNCGNUTVNI-UHFFFAOYSA-N
2,4-dichloro-6-aminophenol,phenol, 2-amino-4,6-dichloro,unii-es86dyt9kh,2-amino-4,6-dichloro-phenol,ccris 7685,es86dyt9kh,6-amino-2,4-dichlorophenol,3,5-dichloro-2-hydroxyaniline,2,4-dichloro-6-amino phenol,acmc-209l1k
10699
2-amino-4,6-dichlorophenol
C1=C(C=C(C(=C1Cl)O)N)Cl
527-62-8
C6H5Cl2NO
178.012
MFCD00035766
WASQBNCGNUTVNI-UHFFFAOYSA-N
2,4-dichloro-6-aminophenol,phenol, 2-amino-4,6-dichloro,unii-es86dyt9kh,2-amino-4,6-dichloro-phenol,ccris 7685,es86dyt9kh,6-amino-2,4-dichlorophenol,3,5-dichloro-2-hydroxyaniline,2,4-dichloro-6-amino phenol,acmc-209l1k
10699
2-amino-4,6-dichlorophenol
C1=C(C=C(C(=C1Cl)O)N)Cl
Color Undesignated
Melting Point 94°C
Quantity 5g
Formula Weight 178.01
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 2-Amino-4,6-dichlorophenol

EINECSNumber : (3)-0990

RTECSNumber : SJ5774000

TSCA : No