Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4,6-dihydroxypyrimidine, 98%

Manufacturer:  Alfa Aesar™ A1181314

Quantity 25g

 View more versions of this product

Catalog No. AAA1181314


  Check Availability
Add to cart

Specifications

Description

Applications
2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.

Solubility
Soluble in Aqueous Alkali.

Notes
Store away from oxidizing agents. Store at -20°C. Store in a cool, dry, well-ventilated area.
Description & Specifications

Chemical Identifiers

56-09-7
127.1
510297
2-Amino-4,6-dihydroxypyrimidine
C4H5N3O2
MFCD00006094
2-Aminopyrimidine-4,6-diol

Specifications

56-09-7
25g
127.1
510297
2-Amino-4,6-dihydroxypyrimidine
>300°C
C4H5N3O2
MFCD00006094
2-Aminopyrimidine-4,6-diol
98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : UW7361000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit