Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,6-dimethylpyrimidine 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
767-15-7
C6H9N3
123.159
MFCD00006102
IDQNBVFPZMCDDN-UHFFFAOYSA-N
2-amino-4,6-dimethylpyrimidine,acetylacetoneguanidine,2-pyrimidinamine, 4,6-dimethyl,4,6-dimethyl-2-pyrimidinamine,4,6-dimethylpyrimidin-2-ylamine,4,6-dimethyl-2-aminopyrimidine,pyrimidine, 2-amino-4,6-dimethyl,unii-4s48mi253m,2-amine-4,6-dimethylpyrimidine,2-amino-4,6-dimethyl pyrimidine
13021
4,6-dimethylpyrimidin-2-amine
CC1=CC(=NC(=N1)N)C
767-15-7
C6H9N3
123.159
MFCD00006102
IDQNBVFPZMCDDN-UHFFFAOYSA-N
2-amino-4,6-dimethylpyrimidine,acetylacetoneguanidine,2-pyrimidinamine, 4,6-dimethyl,4,6-dimethyl-2-pyrimidinamine,4,6-dimethylpyrimidin-2-ylamine,4,6-dimethyl-2-aminopyrimidine,pyrimidine, 2-amino-4,6-dimethyl,unii-4s48mi253m,2-amine-4,6-dimethylpyrimidine,2-amino-4,6-dimethyl pyrimidine
13021
4,6-dimethylpyrimidin-2-amine
CC1=CC(=NC(=N1)N)C
Color White-Yellow
Melting Point 153°C
Quantity 500g
Formula Weight 123.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material 2-Amino-4,6-dimethylpyrimidine