Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-bromophenol, 98%

Manufacturer:  Alfa Aesar™ H5576306

Quantity 5g

 View more versions of this product

Catalog No. AAH5576306


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

40925-68-6
188.02
3236448
C6H6BrNO
MFCD00235171
2-Amino-4-bromophenol

Specifications

40925-68-6
C6H6BrNO
5g
3236448
98%
130° to 135°C
188.02
MFCD00235171
2-Amino-4-bromophenol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit