Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-bromophenol, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
40925-68-6
C6H6BrNO
188.02
JHRIPENGTGSNPJ-UHFFFAOYSA-N
phenol, 2-amino-4-bromo,5-bromo-2-hydroxyaniline,4-bromo-2-aminophenol,2-amino-4-bromo-phenol,2-amino-4-bromophenole,2-amino-4bromophenol,pubchem4511,acmc-209jho,2-amino4-bromophenol,ksc493s8p
351840
2-amino-4-bromophenol
C1=CC(=C(C=C1Br)N)O
40925-68-6
C6H6BrNO
188.02
JHRIPENGTGSNPJ-UHFFFAOYSA-N
phenol, 2-amino-4-bromo,5-bromo-2-hydroxyaniline,4-bromo-2-aminophenol,2-amino-4-bromo-phenol,2-amino-4-bromophenole,2-amino-4bromophenol,pubchem4511,acmc-209jho,2-amino4-bromophenol,ksc493s8p
351840
2-amino-4-bromophenol
C1=CC(=C(C=C1Br)N)O
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 188.02
Percent Purity 98%
Chemical Name or Material 2-Amino-4-bromophenol

GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning