Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Amino-4-bromophenol, 98%, ACROS Organics™
Click to view available options
Quantity:
1g
5g
25g
Specifications
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Quantity | 1g |
Assay Percent Range | 97.5% min. (HPLC) |
Formula Weight | 188.02 |
Percent Purity | 98% |
Chemical Name or Material | 2-Amino-4-bromophenol |
Safety and Handling
GHS H Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning