Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-bromopyrimidine, 98%

Click to view available options
Quantity:
250mg
1g
343926-69-2
C4H4BrN3
174.001
MFCD09999168
MINURGGJJKMQQQ-UHFFFAOYSA-N
2-amino-4-bromopyrimidine,4-bromo-2-pyrimidinamine,2-pyrimidinamine,4-bromo,4-bromanylpyrimidin-2-amine,4-bromopyrimidine-2-ylamine,2-amino-6-bromopyrimidine,4-bromo-2-aminopyrimidine,2-pyrimidinamine,4-bromo-9ci,2-pyrimidinamine, 4-bromo,4-bromo-pyrimidin-2-ylamine
12803496
4-bromopyrimidin-2-amine
C1=CN=C(N=C1Br)N
343926-69-2
C4H4BrN3
174.001
MFCD09999168
MINURGGJJKMQQQ-UHFFFAOYSA-N
2-amino-4-bromopyrimidine,4-bromo-2-pyrimidinamine,2-pyrimidinamine,4-bromo,4-bromanylpyrimidin-2-amine,4-bromopyrimidine-2-ylamine,2-amino-6-bromopyrimidine,4-bromo-2-aminopyrimidine,2-pyrimidinamine,4-bromo-9ci,2-pyrimidinamine, 4-bromo,4-bromo-pyrimidin-2-ylamine
12803496
4-bromopyrimidin-2-amine
C1=CN=C(N=C1Br)N
Applications
It is used as a pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 250mg
Color White
Beilstein 111375
Formula Weight 174
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-4-bromopyrimidine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning