Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-chlorobenzothiazole, 97%

Click to view available options
Quantity:
5g
25g
100g
19952-47-7
C7H5ClN2S
184.641
MFCD00005791
OEQQFQXMCPMEIH-UHFFFAOYSA-N
2-amino-4-chlorobenzothiazole,2-benzothiazolamine, 4-chloro,4-chlorobenzo d thiazol-2-amine,4-chlorobenzothiazol-2-ylamine,benzothiazole, 2-amino-4-chloro,4-chloro-2-benzothiazolamine,4-chloro-2-aminobenzothiazole,4-chloro-benzothiazol-2-ylamine,4-chlorobenzothiazole-2-ylamine,zlchem 166
29872
4-chloro-1,3-benzothiazol-2-amine
C1=CC2=C(C(=C1)Cl)N=C(S2)N
19952-47-7
C7H5ClN2S
184.641
MFCD00005791
OEQQFQXMCPMEIH-UHFFFAOYSA-N
2-amino-4-chlorobenzothiazole,2-benzothiazolamine, 4-chloro,4-chlorobenzo d thiazol-2-amine,4-chlorobenzothiazol-2-ylamine,benzothiazole, 2-amino-4-chloro,4-chloro-2-benzothiazolamine,4-chloro-2-aminobenzothiazole,4-chloro-benzothiazol-2-ylamine,4-chlorobenzothiazole-2-ylamine,zlchem 166
29872
4-chloro-1,3-benzothiazol-2-amine
C1=CC2=C(C(=C1)Cl)N=C(S2)N
Quantity 5g
Melting Point 203°C to 205°C
Beilstein 136784
Formula Weight 184.65
Percent Purity 97%
Chemical Name or Material 2-Amino-4-chlorobenzothiazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 243-439-5

RTECSNumber : DL1225000

TSCA : Yes

Recommended Storage : Ambient temperatures