Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chlorobenzothiazole 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
19952-47-7
C7H5ClN2S
184.641
MFCD00005791
OEQQFQXMCPMEIH-UHFFFAOYSA-N
2-amino-4-chlorobenzothiazole,2-benzothiazolamine, 4-chloro,4-chlorobenzo d thiazol-2-amine,4-chlorobenzothiazol-2-ylamine,benzothiazole, 2-amino-4-chloro,4-chloro-2-benzothiazolamine,4-chloro-2-aminobenzothiazole,4-chloro-benzothiazol-2-ylamine,4-chlorobenzothiazole-2-ylamine,zlchem 166
29872
4-chloro-1,3-benzothiazol-2-amine
C1=CC2=C(C(=C1)Cl)N=C(S2)N
19952-47-7
C7H5ClN2S
184.641
MFCD00005791
OEQQFQXMCPMEIH-UHFFFAOYSA-N
2-amino-4-chlorobenzothiazole,2-benzothiazolamine, 4-chloro,4-chlorobenzo d thiazol-2-amine,4-chlorobenzothiazol-2-ylamine,benzothiazole, 2-amino-4-chloro,4-chloro-2-benzothiazolamine,4-chloro-2-aminobenzothiazole,4-chloro-benzothiazol-2-ylamine,4-chlorobenzothiazole-2-ylamine,zlchem 166
29872
4-chloro-1,3-benzothiazol-2-amine
C1=CC2=C(C(=C1)Cl)N=C(S2)N
Color Yellow
Melting Point 203°C
Quantity 25g
Formula Weight 184.64
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material 2-Amino-4-chlorobenzothiazole

RTECSNumber : DL1225000

TSCA : Yes