Alfa Aesar™ 2-Amino-4-chlorophenol, 98%

Manufacturer: Alfa Aesar™ A1013522

Quantity 100g

 View more versions of this product

Catalog No. AAA1013522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 95-85-2
    Molecular Formula C6H6ClNO
    Formula Weight 143.57
    MDL Number MFCD00007694
    UN Number UN2673
    Beilstein 774859
    Synonym 5-Chloro-2-hydroxyaniline
    Chemical Name or Material 2-Amino-4-chlorophenol

    Specifications

    Formula Weight 143.57
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 95-85-2
    Beilstein 774859
    Synonym 5-Chloro-2-hydroxyaniline
    Quantity 100g
    Molecular Formula C6H6ClNO
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 2-AMINO-4-CHLOROPHENOL
    RTECS Number SJ5700000
    TSCA TSCA
    Melting Point 138° to 142°C
    MDL Number MFCD00007694
    UN Number UN2673
    Chemical Name or Material 2-Amino-4-chlorophenol
    Assay 98%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit