Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-fluorobenzoic acid, 98%

Click to view available options
Quantity:
0.25g
1g
5g
446-32-2
C7H6FNO2
155.128
MFCD00075553
LGPVTNAJFDUWLF-UHFFFAOYSA-N
4-fluoroanthranilic acid,2-amino-4-fluoro-benzoic acid,benzoic acid, 2-amino-4-fluoro,2-amino-4-fluoro benzoic acid,2-amino-4-fluorobenzoicacid,fa0,pubchem1323,amino-4-fluorobenzoic acid,acmc-209jz7,2-amino-4fluorobenzoic acid
2724967
2-amino-4-fluorobenzoic acid
C1=CC(=C(C=C1F)N)C(=O)O
446-32-2
C7H6FNO2
155.128
MFCD00075553
LGPVTNAJFDUWLF-UHFFFAOYSA-N
4-fluoroanthranilic acid,2-amino-4-fluoro-benzoic acid,benzoic acid, 2-amino-4-fluoro,2-amino-4-fluoro benzoic acid,2-amino-4-fluorobenzoicacid,fa0,pubchem1323,amino-4-fluorobenzoic acid,acmc-209jz7,2-amino-4fluorobenzoic acid
2724967
2-amino-4-fluorobenzoic acid
C1=CC(=C(C=C1F)N)C(=O)O
Melting Point 192°C to 196°C
Quantity 5g
Beilstein 2803665
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 2-Amino-4-fluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning