Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-fluoropyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
944401-77-8
C5H5FN2
112.107
MFCD09260906
WCCFLOQQACDOAX-UHFFFAOYSA-N
2-amino-4-fluoropyridine,4-fluoro-2-pyridinamine,4-fluoro-2-pyridylamine,4-fluoro-pyridin-2-ylamine,4-fluoro-2-pyridineamine,amino-4-fluoropyridine,pubchem19911,acmc-209rrv,2-pyridinamine,4-fluoro,4-fluoro-pyridin-2-amine
18327808
4-fluoropyridin-2-amine
C1=CN=C(C=C1F)N
944401-77-8
C5H5FN2
112.107
MFCD09260906
WCCFLOQQACDOAX-UHFFFAOYSA-N
2-amino-4-fluoropyridine,4-fluoro-2-pyridinamine,4-fluoro-2-pyridylamine,4-fluoro-pyridin-2-ylamine,4-fluoro-2-pyridineamine,amino-4-fluoropyridine,pubchem19911,acmc-209rrv,2-pyridinamine,4-fluoro,4-fluoro-pyridin-2-amine
18327808
4-fluoropyridin-2-amine
C1=CN=C(C=C1F)N
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Color Yellow
Quantity 250mg
Beilstein 15519014
Solubility Information Slightly soluble in water.
Formula Weight 112.04
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Amino-4-fluoropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold