Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-hydroxypyridine, 98%

Click to view available options
Quantity:
1g
5g
33631-05-9
C5H6N2O
110.116
MFCD04114161
HQNIMNQVKVPZES-UHFFFAOYSA-N
2-amino-4-hydroxypyridine,2-aminopyridin-4-ol,2-amino-4-pyridinol,4-pyridinol, 2-amino,2-amino-pyridin-4-ol,zlchem 31,pubchem6677,acmc-1acz5,4-hydroxypyridine-2-amine,2-aminopyridin-4 1h-one
820936
2-amino-1H-pyridin-4-one
C1=CNC(=CC1=O)N
33631-05-9
C5H6N2O
110.116
MFCD04114161
HQNIMNQVKVPZES-UHFFFAOYSA-N
2-amino-4-hydroxypyridine,2-aminopyridin-4-ol,2-amino-4-pyridinol,4-pyridinol, 2-amino,2-amino-pyridin-4-ol,zlchem 31,pubchem6677,acmc-1acz5,4-hydroxypyridine-2-amine,2-aminopyridin-4 1h-one
820936
2-amino-1H-pyridin-4-one
C1=CNC(=CC1=O)N
Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Color Yellow
Quantity 5g
Beilstein 385930
Formula Weight 110.12
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Amino-4-hydroxypyridine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C