Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methoxy-6-methyl-1,3,5-triazine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
1668-54-8
C5H8N4O
140.146
MFCD00052764
NXFQWRWXEYTOTK-UHFFFAOYSA-N
2-amino-4-methoxy-6-methyl-1,3,5-triazine,2-methyl-4-amino-6-methoxy-s-triazine,1,3,5-triazin-2-amine, 4-methoxy-6-methyl,2-amino-4-methyl-6-methoxy-1,3,5-triazine,unii-395vj06rc4,s-triazine, 2-amino-4-methoxy-6-methyl,4-methoxy-6-methyl-1,3,5-triazine-2-ylamine,pubchem21415,acmc-1buty,dsstox_cid_21230
15466
CHEBI:83511
4-methoxy-6-methyl-1,3,5-triazin-2-amine
CC1=NC(=NC(=N1)OC)N
1668-54-8
C5H8N4O
140.146
MFCD00052764
NXFQWRWXEYTOTK-UHFFFAOYSA-N
2-amino-4-methoxy-6-methyl-1,3,5-triazine,2-methyl-4-amino-6-methoxy-s-triazine,1,3,5-triazin-2-amine, 4-methoxy-6-methyl,2-amino-4-methyl-6-methoxy-1,3,5-triazine,unii-395vj06rc4,s-triazine, 2-amino-4-methoxy-6-methyl,4-methoxy-6-methyl-1,3,5-triazine-2-ylamine,pubchem21415,acmc-1buty,dsstox_cid_21230
15466
CHEBI:83511
4-methoxy-6-methyl-1,3,5-triazin-2-amine
CC1=NC(=NC(=N1)OC)N
Color Yellow
Quantity 25g
Formula Weight 140.15
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material 2-Amino-4-methoxy-6-methyl-1,3,5-triazine

RTECSNumber : XY3193000

TSCA : Yes