Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4'-methoxyacetophenone hydrochloride, 95%

Click to view available options
Quantity:
1g
5g
3883-94-1
C9H12ClNO2
201.65
MFCD00193078
FZVYWBMMOSHMRS-UHFFFAOYSA-N
2-amino-1-4-methoxyphenyl ethanone hydrochloride,2-amino-4'-methoxyacetophenone hydrochloride,2-amino-1-4-methoxyphenyl ethan-1-one hydrochloride,4-methoxyphenacylamine hydrochloride,ethanone, 2-amino-1-4-methoxyphenyl-, hydrochloride,2-amino-1-4-methoxyphenyl ethan-1-one, chloride,pubchem22350,acmc-1ad40,4-aminoacetyl anisole hydrochloride,2-amino-4-methoxyacetophenone hydrochloride
12487188
2-amino-1-(4-methoxyphenyl)ethanone;hydrochloride
COC1=CC=C(C=C1)C(=O)CN.Cl
3883-94-1
C9H12ClNO2
201.65
MFCD00193078
FZVYWBMMOSHMRS-UHFFFAOYSA-N
2-amino-1-4-methoxyphenyl ethanone hydrochloride,2-amino-4'-methoxyacetophenone hydrochloride,2-amino-1-4-methoxyphenyl ethan-1-one hydrochloride,4-methoxyphenacylamine hydrochloride,ethanone, 2-amino-1-4-methoxyphenyl-, hydrochloride,2-amino-1-4-methoxyphenyl ethan-1-one, chloride,pubchem22350,acmc-1ad40,4-aminoacetyl anisole hydrochloride,2-amino-4-methoxyacetophenone hydrochloride
12487188
2-amino-1-(4-methoxyphenyl)ethanone;hydrochloride
COC1=CC=C(C=C1)C(=O)CN.Cl
Melting Point 190°C to 193°C
Color White
Quantity 1g
Beilstein 3630310
Formula Weight 201.65
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 2-Amino-4'-methoxyacetophenone hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures