Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-methoxypyridine, 95%

Click to view available options
Quantity:
5g
25g
10201-73-7
C6H8N2O
124.143
MFCD07437849
QPHBCOSULYSASF-UHFFFAOYSA-N
2-amino-4-methoxypyridine,4-methoxy-pyridin-2-ylamine,2-amino-4-methoxylpyridine,4-methoxy-2-pyridylamine,2-amino-4-methoxy pyridine,2-pyridinamine, 4-methoxy,zlchem 190,pubchem5700,pubchem16566,4-methoxy-2-pyridinamine
10898715
4-methoxypyridin-2-amine
COC1=CC(=NC=C1)N
10201-73-7
C6H8N2O
124.143
MFCD07437849
QPHBCOSULYSASF-UHFFFAOYSA-N
2-amino-4-methoxypyridine,4-methoxy-pyridin-2-ylamine,2-amino-4-methoxylpyridine,4-methoxy-2-pyridylamine,2-amino-4-methoxy pyridine,2-pyridinamine, 4-methoxy,zlchem 190,pubchem5700,pubchem16566,4-methoxy-2-pyridinamine
10898715
4-methoxypyridin-2-amine
COC1=CC(=NC=C1)N
Applications
2-Amino-4-methoxypyridine is a highly selective inducible nitric oxide synthase inhibitors.

Solubility
Soluble in Dimethyl sulfoxide, methanol. Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Sensitivity Air Sensitive
Quantity 25g
Color Yellow
Melting Point 115°C to 118°C
UN Number UN2811
Beilstein 471837
Formula Weight 124.14
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Amino-4-methoxypyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.