Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methyl-5-acetylthiazole, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
30748-47-1
C6H8N2OS
156.203
MFCD00051952
PKUKCASRNJIQNU-UHFFFAOYSA-N
2-amino-4-methyl-5-acetylthiazole,1-2-amino-4-methylthiazol-5-yl ethanone,1-2-amino-4-methyl-1,3-thiazol-5-yl ethanone,5-acetyl-2-amino-4-methylthiazole,1-2-amino-4-methyl-1,3-thiazol-5-yl ethan-1-one,1-2-amino-4-methyl-thiazol-5-yl-ethanone,5-acetyl-2-amino-4-methyl-1,3-thiazole,ethanone, 1-2-amino-4-methyl-5-thiazolyl,1-2-amino-4-methyl-1,3-thiazol-5-yl-1-ethanone,5-acetyl-4-methyl-2-thiazolamine
720882
1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethanone
CC1=C(SC(=N1)N)C(=O)C
30748-47-1
C6H8N2OS
156.203
MFCD00051952
PKUKCASRNJIQNU-UHFFFAOYSA-N
2-amino-4-methyl-5-acetylthiazole,1-2-amino-4-methylthiazol-5-yl ethanone,1-2-amino-4-methyl-1,3-thiazol-5-yl ethanone,5-acetyl-2-amino-4-methylthiazole,1-2-amino-4-methyl-1,3-thiazol-5-yl ethan-1-one,1-2-amino-4-methyl-thiazol-5-yl-ethanone,5-acetyl-2-amino-4-methyl-1,3-thiazole,ethanone, 1-2-amino-4-methyl-5-thiazolyl,1-2-amino-4-methyl-1,3-thiazol-5-yl-1-ethanone,5-acetyl-4-methyl-2-thiazolamine
720882
1-(2-amino-4-methyl-1,3-thiazol-5-yl)ethanone
CC1=C(SC(=N1)N)C(=O)C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 98%
Formula Weight 156.21
Chemical Name or Material 2-Amino-4-methyl-5-acetylthiazole

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning