Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methylbenzonitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7058791
Click to view available options
Quantity:
1 g
25 g
26830-96-6
C8H8N2
132.17
MFCD00173706
LGNVAEIITHYWCG-UHFFFAOYSA-N
2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
2801276
2-amino-4-methylbenzonitrile
CC1=CC=C(C#N)C(N)=C1
26830-96-6
C8H8N2
132.17
MFCD00173706
LGNVAEIITHYWCG-UHFFFAOYSA-N
2-cyano-5-methylaniline, 3-amino-4-cyanotoluene, 2-amino-4-methyl-benzonitrile, 4-methyl-2-aminobenzonitrile, benzonitrile, 2-amino-4-methyl, 2-amino-4-methylbenzenecarbonitrile, 2-amino-4-methylbenzonitril, 6-cyano-m-toluidine, pubchem4661, 4-methylanthranilonitrile
2801276
2-amino-4-methylbenzonitrile
CC1=CC=C(C#N)C(N)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 91°C to 93°C
Quantity 1 g
UN Number UN3439
Formula Weight 132.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-4-methylbenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 248-020-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit