Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-methylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
863578-36-3
C13H20BNO2
233.12
PEXRISJTWBBZRY-UHFFFAOYSA-N
2-amino-4-methylphenylboronic acid, pinacol ester,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,2-amino-4-methylphenylboronic acid pinacol ester,pexrisjtwbbzry-uhfffaoysa-n,2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5-methylphenylamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine,benzenamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,5-methyl-2--4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
17750209
863578-36-3
C13H20BNO2
233.12
PEXRISJTWBBZRY-UHFFFAOYSA-N
2-amino-4-methylphenylboronic acid, pinacol ester,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,2-amino-4-methylphenylboronic acid pinacol ester,pexrisjtwbbzry-uhfffaoysa-n,2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5-methylphenylamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenylamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylamine,benzenamine,5-methyl-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl,5-methyl-2--4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline
17750209
Melting Point 48°C to 50°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 233.12
Percent Purity 97%
Chemical Name or Material 2-Amino-4-methylphenylboronic acid pinacol ester

GHS Signal Word: Warning