Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-4-nitrobenzoic acid, ≥97%

Click to view available options
Quantity:
25g
100g
500g
619-17-0
C7H6N2O4
182.135
MFCD00007262
UEALKTCRMBVTFN-UHFFFAOYSA-N
2-amino-4-nitrobenzoic acid,4-nitroanthranilic acid,benzoic acid, 2-amino-4-nitro,2-amino-4-nitrobenzoicacid,anthranilic acid, 4-nitro,ccris 441,2-amino-4-nitro-benzoic acid,4-nitro-2-aminobenzoic acid,dsstox_cid_963,dsstox_rid_75891
12076
CHEBI:75335
C1=CC(=C(C=C1[N+](=O)[O-])N)C(=O)O
619-17-0
C7H6N2O4
182.135
MFCD00007262
UEALKTCRMBVTFN-UHFFFAOYSA-N
2-amino-4-nitrobenzoic acid,4-nitroanthranilic acid,benzoic acid, 2-amino-4-nitro,2-amino-4-nitrobenzoicacid,anthranilic acid, 4-nitro,ccris 441,2-amino-4-nitro-benzoic acid,4-nitro-2-aminobenzoic acid,dsstox_cid_963,dsstox_rid_75891
12076
CHEBI:75335
C1=CC(=C(C=C1[N+](=O)[O-])N)C(=O)O
Quantity 500g
Melting Point 257°C (literature)
Beilstein 785071
Formula Weight 182.13
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material 2-Amino-4-nitrobenzoic acid

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CB3675000

TSCA : TSCA