Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-amino-4-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
695-34-1
C6H8N2
108.14
MFCD00006321
ORLGLBZRQYOWNA-UHFFFAOYSA-N
2-amino-4-methylpyridine,2-amino-4-picoline,2-pyridinamine, 4-methyl,ascensil,4-methyl-2-pyridinamine,4-methyl-2-pyridylamine,4-methyl-2-aminopyridine,4-picoline, 2-amino,w 45 raschig,aminton
1533
4-methylpyridin-2-amine
CC1=CC(=NC=C1)N
695-34-1
C6H8N2
108.14
MFCD00006321
ORLGLBZRQYOWNA-UHFFFAOYSA-N
2-amino-4-methylpyridine,2-amino-4-picoline,2-pyridinamine, 4-methyl,ascensil,4-methyl-2-pyridinamine,4-methyl-2-pyridylamine,4-methyl-2-aminopyridine,4-picoline, 2-amino,w 45 raschig,aminton
1533
4-methylpyridin-2-amine
CC1=CC(=NC=C1)N
Flash Point 118°C
Boiling Point 230°C
Melting Point 96°C to 101°C
Water 0.5% max.
Infrared Spectrum Authentic
Quantity 500g
Packaging Glass bottle
Color Beige to Yellow or Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 22, V,9, 325
Merck Index 15, 462
Solubility Information Solubility in water: 41g/L (20°C). Other solubilities: soluble in lower alcohols,slightly soluble in petroleum ether
Formula Weight 108.14
Physical Form Flakes
Percent Purity 98%
Chemical Name or Material 2-Amino-4-picoline
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.
May cause respiratory irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 211-780-9