Alfa Aesar™ 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H54372MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH54372MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 944401-57-4
    Molecular Formula C12H16BF3N2O2
    Formula Weight 288.07
    MDL Number MFCD12923420
    Beilstein 15519102
    Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine
    Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester

    Specifications

    Formula Weight 288.07
    Molecular Formula C12H16BF3N2O2
    MDL Number MFCD12923420
    CAS 944401-57-4
    Quantity 250mg
    Beilstein 15519102
    Synonym 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
    Assay 95%

    Description

    Applications
    2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit