Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-(trifluoromethyl)pyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
500mg
106447-97-6
C6H5F3N2
162.11
RWGBXAQMUBGGKQ-UHFFFAOYSA-N
2-amino-4-trifluoromethyl pyridine,4-trifluoromethyl pyridin-2-amine,4-trifluoromethyl-pyridin-2-ylamine,2-amino-4-trifluoromethylpyridine,4-trifluoromethyl-2-pyridinamine,4-trifluoromethyl-2-pyridylamine,2-pyridinamine, 4-trifluoromethyl,2-amino-4-trifluoromethyl-pyridine,pubchem2977
2778315
4-(trifluoromethyl)pyridin-2-amine
C1=CN=C(C=C1C(F)(F)F)N
106447-97-6
C6H5F3N2
162.11
RWGBXAQMUBGGKQ-UHFFFAOYSA-N
2-amino-4-trifluoromethyl pyridine,4-trifluoromethyl pyridin-2-amine,4-trifluoromethyl-pyridin-2-ylamine,2-amino-4-trifluoromethylpyridine,4-trifluoromethyl-2-pyridinamine,4-trifluoromethyl-2-pyridylamine,2-pyridinamine, 4-trifluoromethyl,2-amino-4-trifluoromethyl-pyridine,pubchem2977
2778315
4-(trifluoromethyl)pyridin-2-amine
C1=CN=C(C=C1C(F)(F)F)N
Melting Point 73.5°C to 75.5°C
Color Brown to White
Quantity 100mg
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 162.11
Physical Form Powder, Solid or Crystalline
Percent Purity 97%
Chemical Name or Material 2-Amino-4-(trifluoromethyl)pyridine
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning