Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromo-3-picoline, 97%, Thermo Scientific™

Catalog Number AC360880100
Click to view available options
Quantity:
10 g
25 g
3430-21-5
C6H7BrN2
187.04
KBLGGRWUEVCNPY-UHFFFAOYSA-N
2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
137934
5-bromo-3-methylpyridin-2-amine
CC1=CC(=CN=C1N)Br
3430-21-5
C6H7BrN2
187.04
KBLGGRWUEVCNPY-UHFFFAOYSA-N
2-amino-5-bromo-3-methylpyridine, 2-amino-5-bromo-3-picoline, 2-amino-3-methyl-5-bromopyridine, 5-bromo-3-methyl-pyridin-2-ylamine, 2-pyridinamine, 5-bromo-3-methyl, 5-bromo-3-methyl-2-pyridinamine, 2-amina-5-bromo-3-picoline, 5-bromo-3-methyl-2-pyridylamine, 5-bromo-3-methyl-pyridin-2-amine, 5-bromo-3-methylpyridin-2-ylamine
137934
5-bromo-3-methylpyridin-2-amine
CC1=CC(=CN=C1N)Br
Melting Point 92.0°C to 94.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 10 g
Solubility Information Solubility in water: slighlty soluble. Other solubilities: soluble in methanol
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3-picoline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit