Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-bromo-3-picoline, 97%, Acros Organics™

Click to view available options
Quantity:
10g
25g
3430-21-5
C6H7BrN2
187.04
KBLGGRWUEVCNPY-UHFFFAOYSA-N
2-amino-5-bromo-3-methylpyridine,2-amino-5-bromo-3-picoline,2-amino-3-methyl-5-bromopyridine,5-bromo-3-methyl-pyridin-2-ylamine,2-pyridinamine, 5-bromo-3-methyl,5-bromo-3-methyl-2-pyridinamine,2-amina-5-bromo-3-picoline,5-bromo-3-methyl-2-pyridylamine,5-bromo-3-methyl-pyridin-2-amine,5-bromo-3-methylpyridin-2-ylamine
137934
5-bromo-3-methylpyridin-2-amine
CC1=CC(=CN=C1N)Br
3430-21-5
C6H7BrN2
187.04
KBLGGRWUEVCNPY-UHFFFAOYSA-N
2-amino-5-bromo-3-methylpyridine,2-amino-5-bromo-3-picoline,2-amino-3-methyl-5-bromopyridine,5-bromo-3-methyl-pyridin-2-ylamine,2-pyridinamine, 5-bromo-3-methyl,5-bromo-3-methyl-2-pyridinamine,2-amina-5-bromo-3-picoline,5-bromo-3-methyl-2-pyridylamine,5-bromo-3-methyl-pyridin-2-amine,5-bromo-3-methylpyridin-2-ylamine
137934
5-bromo-3-methylpyridin-2-amine
CC1=CC(=CN=C1N)Br
Quantity 25g
Melting Point 92.0°C to 94.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: slighlty soluble. Other solubilities: soluble in methanol
Formula Weight 187.04
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3-picoline

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning