Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-5-bromo-3-pyridinemethanol, ≥98%

Click to view available options
Quantity:
1g
5g
335031-01-1
C6H7BrN2O
203.039
MFCD05664025
HIVQAWXDBPYNDU-UHFFFAOYSA-N
2-amino-5-bromo-3-hydroxymethyl pyridine,2-amino-5-bromopyridin-3-yl methanol,2-amino-5-bromo-3-pyridinemethanol,2-amino-5-bromopyridine-3-methanol,2-amino-5-bromo-3-pyridinyl methanol,2-amino-5-bromo-pyridin-3-yl-methanol,3-pyridinemethanol, 2-amino-5-bromo,2-amino-5-bromo-3-pyridyl methan-1-ol,2-amino-5-bromopyridin-3-yl-methanol,pubchem9344
10891366
(2-amino-5-bromopyridin-3-yl)methanol
C1=C(C(=NC=C1Br)N)CO
335031-01-1
C6H7BrN2O
203.039
MFCD05664025
HIVQAWXDBPYNDU-UHFFFAOYSA-N
2-amino-5-bromo-3-hydroxymethyl pyridine,2-amino-5-bromopyridin-3-yl methanol,2-amino-5-bromo-3-pyridinemethanol,2-amino-5-bromopyridine-3-methanol,2-amino-5-bromo-3-pyridinyl methanol,2-amino-5-bromo-pyridin-3-yl-methanol,3-pyridinemethanol, 2-amino-5-bromo,2-amino-5-bromo-3-pyridyl methan-1-ol,2-amino-5-bromopyridin-3-yl-methanol,pubchem9344
10891366
(2-amino-5-bromopyridin-3-yl)methanol
C1=C(C(=NC=C1Br)N)CO
Quantity 5g
Color White to Yellow
Beilstein 9387621
Formula Weight 203.04
Physical Form Solid
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-bromo-3-pyridinemethanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C