Learn More
Accela Chembio Inc 2-amino-5-bromo-4-cyclopropylthiazole | 1g | 869854-12-6 | MFCD13193636 | 95+% | Shelf Life: 180 Days | Light Sensitive/air Sensitive/+4
SDP
Supplier:  Accela Chembio Inc SY0110641G
Encompass_Preferred
2-amino-5-bromo-4-cyclopropylthiazole | 1g | 869854-12-6 | MFCD13193636 | 95+% | Shelf Life: 180 Days | Light Sensitive/air Sensitive/+4
Catalog No. 50-251-672