Alfa Aesar™ 2-Amino-5-bromo-4-methylpyridine, 97%

Manufacturer: Alfa Aesar™ 4462314

Quantity 25g

 View more versions of this product

Catalog No. AA4462314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 98198-48-2
    Molecular Formula C6H7BrN2
    Formula Weight 187.04
    MDL Number MFCD03427660
    Synonym 2-Amino-5-bromo-4-picoline; 5-Bromo-4-methyl-2-pyridinamine
    Chemical Name or Material 2-Amino-5-bromo-4-methylpyridine

    Specifications

    Formula Weight 187.04
    Synonym 2-Amino-5-bromo-4-picoline; 5-Bromo-4-methyl-2-pyridinamine
    Color Cream
    MDL Number MFCD03427660
    CAS 98198-48-2
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Physical Form Powder
    Melting Point 148° to 151°C
    Molecular Formula C6H7BrN2
    Quantity 25g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-Amino-5-bromo-4-methylpyridine
    Assay 97%

    GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit